missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrochalcone, 97%

Catalog No. AAA1776314
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1776314 25 g
AAA1776306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1776314 Supplier Thermo Scientific Chemicals Supplier No. A1776314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7473-93-0 | C15H11NO3 | 253.257 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7473-93-0
Molecular Formula C15H11NO3
Molecular Weight (g/mol) 253.257
MDL Number MFCD00031069
InChI Key KTXHLWZQKQDFRF-ZHACJKMWSA-N
Synonym 2-nitrochalcone, chalcone, 2-nitro, 2-nitrobenzylideneacetophenone, 2e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, ccris 1671, e-3-2-nitro-phenyl-1-phenyl-propenone, 2-propen-1-one, 3-2-nitrophenyl-1-phenyl, e-3-2-nitrophenyl-1-phenyl-2-propen-1-one, e-3-2-nitrophenyl-1-phenylprop-2-en-1-one, e-3-2-nitrophenyl-1-phenyl-prop-2-en-1-one
PubChem CID 5337611
IUPAC Name (E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2[N+](=O)[O-]

Specifications

Melting Point 124°C to 128°C
Quantity 25 g
Formula Weight 253.26
Percent Purity 97%
Chemical Name or Material 2-Nitrochalcone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only