CAS RN 7473-93-0
IUPAC Name:
(2E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
Synonyms:
(2E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one
Molecular Weight (g/mol):
253.26
Molecular Formula:
C15H11NO3
InChi Key:
KTXHLWZQKQDFRF-ZHACJKMWSA-N
SMILES:
[O-][N+](=O)C1=CC=CC=C1\C=C\C(=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
2-Nitrochalcone, 97%
2-Nitrochalcone, 97%
CAS: 7473-93-0 Molecular Formula: C15H11NO3 Molecular Weight (g/mol): 253.257 MDL Number: MFCD00031069 InChI Key: KTXHLWZQKQDFRF-ZHACJKMWSA-N Synonym: 2-nitrochalcone,chalcone, 2-nitro,2-nitrobenzylideneacetophenone,2e-3-2-nitrophenyl-1-phenylprop-2-en-1-one,ccris 1671,e-3-2-nitro-phenyl-1-phenyl-propenone,2-propen-1-one, 3-2-nitrophenyl-1-phenyl,e-3-2-nitrophenyl-1-phenyl-2-propen-1-one,e-3-2-nitrophenyl-1-phenylprop-2-en-1-one,e-3-2-nitrophenyl-1-phenyl-prop-2-en-1-one PubChem CID: 5337611 IUPAC Name: (E)-3-(2-nitrophenyl)-1-phenylprop-2-en-1-one SMILES: C1=CC=C(C=C1)C(=O)C=CC2=CC=CC=C2[N+](=O)[O-]