missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobiphenyl 98.0+%, TCI America™

Catalog No. p-7133672
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N019425G 25 g
N0194500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N019425G Supplier TCI America Supplier No. N019425G

Chemical Identifiers

CAS 86-00-0
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD00007126
InChI Key YOJKKXRJMXIKSR-UHFFFAOYSA-N
Synonym 2-nitrobiphenyl, 2-nitro-1,1'-biphenyl, 2-nitrodiphenyl, o-nitrobiphenyl, o-nitrodiphenyl, biphenyl, 2-nitro, 1,1'-biphenyl, 2-nitro, nitrobiphenyl, 2-phenylnitrobenzene, 2-nitro-biphenyl
PubChem CID 6829
IUPAC Name 2-nitro-1,1'-biphenyl
SMILES [O-][N+](=O)C1=CC=CC=C1C1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 199.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Nitrobiphenyl
RTECSNumber DV5530000
TSCA No