missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzyl Cyanide 98.0+%, TCI America™
$179.05 - $179.05
Chemical Identifiers
| CAS | 610-66-2 |
|---|---|
| Molecular Formula | C8H6N2O2 |
| Molecular Weight (g/mol) | 162.15 |
| MDL Number | MFCD00007183 |
| InChI Key | YPRFCQAWSNWRLM-UHFFFAOYSA-N |
| Synonym | 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl |
| PubChem CID | 11888 |
| IUPAC Name | 2-(2-nitrophenyl)acetonitrile |
| SMILES | [O-][N+](=O)C1=CC=CC=C1CC#N |
Chemical Identifiers
| 610-66-2 | |
| 162.15 | |
| YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
| 11888 | |
| [O-][N+](=O)C1=CC=CC=C1CC#N |
| C8H6N2O2 | |
| MFCD00007183 | |
| 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl | |
| 2-(2-nitrophenyl)acetonitrile |
Specifications
| 610-66-2 | |
| 178°C | |
| MFCD00007183 | |
| 3439 | |
| YPRFCQAWSNWRLM-UHFFFAOYSA-N | |
| 2-(2-nitrophenyl)acetonitrile | |
| 11888 | |
| ≥98.0% (GC) | |
| 2-Nitrobenzyl Cyanide |
| 84°C | |
| C8H6N2O2 | |
| 5 g | |
| 2-nitrophenylacetonitrile, 2-2-nitrophenyl acetonitrile, 2-nitrophenyl acetonitrile, benzeneacetonitrile, 2-nitro, 2-nitrobenzyl cyanide, o-nitrophenyl acetonitrile, o-nitrobenzacetonitrile, 2-nitrobenzeneacetonitrile, 2-nitrobenzyl nitrile, acetonitrile, o-nitrophenyl | |
| [O-][N+](=O)C1=CC=CC=C1CC#N | |
| 162.15 | |
| 162.15 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-1820
RTECSNumber : AM1248000
TSCA : Yes