missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl Alcohol 98.0+%, TCI America™

Catalog No. N017925G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N017925G 25 g
N0179250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N017925G Supplier TCI America Supplier No. N017925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 612-25-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007186
InChI Key BWRBVBFLFQKBPT-UHFFFAOYSA-N
Synonym 2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
PubChem CID 11923
IUPAC Name (2-nitrophenyl)methanol
SMILES OCC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 74°C
Color Yellow
Boiling Point 168°C
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzyl Alcohol
RTECSNumber DP0657050
TSCA No