missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzyl alcohol, 97%

Catalog No. p-7022628
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1549614 25 g
AAA1549622 100 g
AAA1549636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1549614 Supplier Thermo Scientific Chemicals Supplier No. A1549614

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 612-25-9 | C7H7NO3 | 153.14 g/mol

The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The o-nitrobenzyl group (o-NB) is utilized frequently in polymer and materials science. It covers the use of based cross-linkers for photodegradable hydrogels, o-NB side chain functionalization in (block) copolymers, o-NB side chain functionalization for thin film patterning, o-NB for self-assembled monolayers, photo cleavable block copolymers, and photo cleavable bioconjugates.

Solubility
Soluble in water 5000 mg/L 20 deg Celsius.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 612-25-9
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007186
InChI Key BWRBVBFLFQKBPT-UHFFFAOYSA-N
Synonym 2-nitrobenzyl alcohol, 2-nitrophenyl methanol, o-nitrobenzyl alcohol, benzyl alcohol, o-nitro, benzenemethanol, 2-nitro, 2-nitrobenzenemethanol, o-hydroxymethylnitrobenzene, ccris 7967, benzenemethanol, nitro, 2-nitrophenyl methan-1-ol
PubChem CID 11923
IUPAC Name (2-nitrophenyl)methanol
SMILES OCC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 70°C to 74°C
Boiling Point 270°C
Quantity 25 g
Beilstein 2046649
Sensitivity Light sensitive
Solubility Information Soluble in water 5000mg/L 20 deg Celsius.
Formula Weight 153.14
Percent Purity 97%
Chemical Name or Material 2-Nitrobenzyl alcohol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 210-302-6
RTECSNumber DP0657050
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only