missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoic Acid 90.0+%, TCI America™

Catalog No. p-7133650
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N015525G 25 g
N0155500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N015525G Supplier TCI America Supplier No. N015525G
Only null left

Chemical Identifiers

CAS 552-16-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007137
InChI Key SLAMLWHELXOEJZ-UHFFFAOYSA-N
Synonym 2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
PubChem CID 11087
ChEBI CHEBI:25620
SMILES C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 144°C
Color Yellow
Quantity 25 g
Formula Weight 167.12
Percent Purity ≥90.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzoic Acid
EINECSNumber (3)-1505
RTECSNumber DH5050000
TSCA Yes