missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzoic acid, 95%

Catalog No. AAA1039322
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1039322 100 g
AAA1039336 500 g
AAA103930E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1039322 Supplier Thermo Scientific Chemicals Supplier No. A1039322
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 552-16-9 | C7H5NO4 | 167.12 g/mol

It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a synthetic reagent and can function as a growth supplement for Pseudomonas fluorescens strain KU-7. It also finds its use as a reagent for protection of amine groups and in organic synthesis.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 552-16-9
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007137
InChI Key SLAMLWHELXOEJZ-UHFFFAOYSA-N
Synonym 2-nitrobenzoic acid, o-nitrobenzoic acid, benzoic acid, 2-nitro, 2-nitrobenzoicacid, o-carboxynitrobenzene, benzoic acid, o-nitro, nitrobenzoic acid, 2-nitro-benzoic acid, ccris 2334, 2-nitro benzoic acid
PubChem CID 11087
ChEBI CHEBI:25620
SMILES C1=CC=C(C(=C1)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 146°C to 149°C
Density 1.58
Quantity 100 g
Beilstein 1910632
Merck Index 14,6589
Solubility Information Soluble in water.
Formula Weight 167.12
Percent Purity 95%
Chemical Name or Material 2-Nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H341-H315-H319-H335
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-004-9
RTECSNumber DH5050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only