missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzhydrazide 98.0+%, TCI America™

Catalog No. N014825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N014825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N014825G Supplier TCI America Supplier No. N014825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 606-26-8
Molecular Formula C7H7N3O3
Molecular Weight (g/mol) 181.151
MDL Number MFCD00007598
InChI Key LYGGDXLOJMNFBV-UHFFFAOYSA-N
Synonym 2-nitrobenzhydrazide, o-nitrobenzohydrazide, o-nitrobenzhydrazide, 2-nitro-benzoic acid hydrazide, 2-nitrobenzoic hydrazide, benzoic acid, 2-nitro-, hydrazide, benzoylhydrazine, o-nitro, 2-nitrobenzenecarbohydrazide, nitrobenzhydrazid, 2-nitrobenzoylhydrazine
PubChem CID 3781218
IUPAC Name 2-nitrobenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)[N+](=O)[O-]

Specifications

Melting Point 120°C
Color Yellow
Quantity 25 g
Formula Weight 181.15
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzhydrazide