missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Nitrobenzenesulfonamide 98.0+%, TCI America™
Supplier: TCI America N070525G
Specifications
| 2-Nitrobenzenesulfonamide | |
| 191°C | |
| MFCD00009807 | |
| 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n | |
| NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O | |
| 202.18 | |
| 202.18 | |
| Crystalline Powder |
| 5455-59-4 | |
| C6H6N2O4S | |
| 25 g | |
| GNDKYAWHEKZHPJ-UHFFFAOYSA-N | |
| 2-nitrobenzene-1-sulfonamide | |
| 138510 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 5455-59-4 | |
| 202.18 | |
| GNDKYAWHEKZHPJ-UHFFFAOYSA-N | |
| 138510 | |
| NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O |
| C6H6N2O4S | |
| MFCD00009807 | |
| 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n | |
| 2-nitrobenzene-1-sulfonamide |
Safety and Handling
TSCA : Yes