missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonamide 98.0+%, TCI America™

Catalog No. N07055G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07055G 5 g
N070525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07055G Supplier TCI America Supplier No. N07055G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5455-59-4
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00009807
InChI Key GNDKYAWHEKZHPJ-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n
PubChem CID 138510
IUPAC Name 2-nitrobenzene-1-sulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 191°C
Quantity 5 g
Formula Weight 202.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzenesulfonamide