missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfonamide, 97+%

Catalog No. AAA1568922
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1568922 100 g
AAA1568906 5 g
AAA1568909 10 g
AAA1568914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AAA1568922 Supplier Thermo Scientific Chemicals Supplier No. A1568922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5455-59-4 | C6H6N2O4S | 202.18 g/mol

It is applied as a reagent for synthesis of medium-ring amines via N-alkylation, e.g. with ω-bromo alkanols under Mitsunobu conditions (DEAD/PPh3), and cyclization of the resulting ω-bromo alkylsulfonamides (Cs2CO3/Bu4NI). It is a carbonic anhydrase inhibitor. It is employed as a reactant involved in synthesis of cyclic nitrogen compounds via intramolecular hydroamination, pentacyclic lycopodium alkaloid huperzine-Q2, pyrrolidines, intermolecular C-H insertion reactions and reactant involved in intermolecular amination of allyl alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as a reagent for synthesis of medium-ring amines via N-alkylation, e.g. with ω-bromo alkanols under Mitsunobu conditions (DEAD/PPh3), and cyclization of the resulting ω-bromo alkylsulfonamides (Cs2CO3/Bu4NI). It is a carbonic anhydrase inhibitor. It is employed as a reactant involved in synthesis of cyclic nitrogen compounds via intramolecular hydroamination, pentacyclic lycopodium alkaloid huperzine-Q2, pyrrolidines, intermolecular C-H insertion reactions and reactant involved in intermolecular amination of allyl alcohols.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 5455-59-4
Molecular Formula C6H6N2O4S
Molecular Weight (g/mol) 202.18
MDL Number MFCD00009807
InChI Key GNDKYAWHEKZHPJ-UHFFFAOYSA-N
Synonym 2-nitrobenzene-1-sulfonamide, 2-nitrobenzenesulphonamide, 2-nitro-benzenesulfonamide, nitrobenzenesulfonamide, 2-nitrophenylsulfonamide, 2-sulphamoylnitrobenzene, acmc-209lhg, 2-nitrobenzene-sulfonamide, 2-nitro benzene sulphonamide, ksc490i0n
PubChem CID 138510
IUPAC Name 2-nitrobenzenesulfonamide
SMILES NS(=O)(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 188°C to 194°C
Quantity 100 g
Beilstein 2214215
Solubility Information Slightly soluble in water.
Formula Weight 202.19
Percent Purity ≥98%
Chemical Name or Material 2-Nitrobenzenesulfonamide
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only