missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfenyl chloride, 98%

Catalog No. AC161330100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC161330100 10 g Glass bottle
AC161330500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC161330100 Supplier Thermo Scientific Chemicals Supplier No. 161330100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7669-54-7 | C6H4ClNO2S | 189.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7669-54-7
Molecular Formula C6H4ClNO2S
Molecular Weight (g/mol) 189.62
MDL Number MFCD00007128
InChI Key NTNKNFHIAFDCSJ-UHFFFAOYSA-N
Synonym 2-nitrobenzenesulfenyl chloride, 2-nitrophenylsulfenyl chloride, benzenesulfenyl chloride, 2-nitro, o-nitrobenzenesulfenyl chloride, o-nitrophenylsulfenyl chloride, 2-nitrobenzenesulfenylchloride, o-nitrophenylsulphenyl chloride, o-nitrobenzenesulphenyl chloride, 2-nitrophenyl thiohypochlorite, 2-nitrophenyl sulfanyl chlorane
PubChem CID 24319
IUPAC Name (2-nitrophenyl) thiohypochlorite
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])SCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 70.0°C to 75.0°C
Color Golden to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula O2NC6H4SCl
Quantity 10 g
Beilstein 06, I, 157
Fieser 01,745; 04,359; 06,419; 08,360; 10,277; 15,217
Solubility Information Solubility in water: decomposes
Formula Weight 189.62
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2-Nitrobenzenesulfenyl chloride, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 231-644-2
TSCA TSCA

RUO – Research Use Only