missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzenesulfenyl chloride, 97%

Catalog No. AAB2362709
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2362709 10 g
AAB2362718 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2362709 Supplier Thermo Scientific Chemicals Supplier No. B2362709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 7669-54-7 | C6H4ClNO2S | 189.613 g/mol

2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitrobenzenesulfenyl chloride is used as a reagent for N-protection as N-2-nitrophenylsulfenyl (Nps) derivatives, in amino acids and peptides and nucleosides.

Solubility
Reacts with water.

Notes
Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.

Chemical Identifiers

CAS 7669-54-7
Molecular Formula C6H4ClNO2S
Molecular Weight (g/mol) 189.613
MDL Number MFCD00007128
InChI Key NTNKNFHIAFDCSJ-UHFFFAOYSA-N
Synonym 2-nitrobenzenesulfenyl chloride, 2-nitrophenylsulfenyl chloride, benzenesulfenyl chloride, 2-nitro, o-nitrobenzenesulfenyl chloride, o-nitrophenylsulfenyl chloride, 2-nitrobenzenesulfenylchloride, o-nitrophenylsulphenyl chloride, o-nitrobenzenesulphenyl chloride, 2-nitrophenyl thiohypochlorite, 2-nitrophenyl sulfanyl chlorane
PubChem CID 24319
IUPAC Name (2-nitrophenyl) thiohypochlorite
SMILES C1=CC=C(C(=C1)[N+](=O)[O-])SCl

Specifications

Melting Point 72°C to 75°C
Quantity 10 g
UN Number UN3261
Beilstein 473741
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 189.62
Percent Purity 98%
Chemical Name or Material 2-Nitrobenzenesulfenyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 231-644-2
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only