missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzeneboronic acid pinacol ester, 98+%

CAS: 190788-59-1 | C12H16BNO4 | 249.07 g/mol

Supplier:  Thermo Scientific Chemicals L1996306

 View more versions of this product

Catalog No. AAL1996306


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Nitrobenzeneboronic acid pinacol ester
42°C to 44°C
MFCD02179447
7708824
VLJYUDGCEKORNG-UHFFFAOYSA-N
4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
3699568
≥98%
190788-59-1
C12H16BNO4
5 g
4,4,5,5-tetramethyl-2-2-nitrophenyl-1,3,2-dioxaborolane, 2-nitrobenzeneboronic acid pinacol ester, 2-nitrophenyl boronic acid pinacol ester, 2-nitrophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-nitrophenyl, 2-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, amtb916, 2-nitrophenyl boronic acid, pinacol ester, 2-nitrophenylboronic acid, pinacol ester
CC1(C)OB(OC1(C)C)C1=CC=CC=C1[N+]([O-])=O
249.07
249.08

Chemical Identifiers

190788-59-1
249.07
VLJYUDGCEKORNG-UHFFFAOYSA-N
3699568
CC1(C)OB(OC1(C)C)C1=CC=CC=C1[N+]([O-])=O
C12H16BNO4
MFCD02179447
4,4,5,5-tetramethyl-2-2-nitrophenyl-1,3,2-dioxaborolane, 2-nitrobenzeneboronic acid pinacol ester, 2-nitrophenyl boronic acid pinacol ester, 2-nitrophenylboronic acid pinacol ester, 1,3,2-dioxaborolane, 4,4,5,5-tetramethyl-2-2-nitrophenyl, 2-2-nitrophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl nitrobenzene, amtb916, 2-nitrophenyl boronic acid, pinacol ester, 2-nitrophenylboronic acid, pinacol ester
4,4,5,5-tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only