missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzaldehyde 99.0+%, TCI America™

Catalog No. p-7133637
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N013025G 25 g
N0130100G 100 g
N0130500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. N013025G Supplier TCI America Supplier No. N013025G
Only null left

Chemical Identifiers

CAS 552-89-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.121
MDL Number MFCD00007132
InChI Key CMWKITSNTDAEDT-UHFFFAOYSA-N
Synonym o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
PubChem CID 11101
ChEBI CHEBI:66927
IUPAC Name 2-nitrobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)[N+](=O)[O-]

Specifications

Melting Point 44°C
Boiling Point 236°C
Quantity 25 g
Formula Weight 151.12
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitrobenzaldehyde
EINECSNumber (3)-1181
RTECSNumber CU7300000
TSCA Yes