missing translation for 'onlineSavingsMsg'
Learn More

2-Nitrobenzaldehyde, 99+%

Catalog No. p-4509457
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC128400250 25 g Plastic bottle
AC128401000 100 g Plastic bottle
AC128405000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC128400250 Supplier Thermo Scientific Chemicals Supplier No. 128400250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 552-89-6 | C7H5NO3 | 151.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-89-6
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
InChI Key CMWKITSNTDAEDT-UHFFFAOYSA-N
Synonym o-nitrobenzaldehyde, benzaldehyde, 2-nitro, nitrobenzaldehyde, benzaldehyde, o-nitro, 2-nitro-benzaldehyde, 2-formylnitrobenzene, ccris 2322, unii-48b18q9b8e, benzaldehyde, nitro, ortho-nitrobenzaldehyde
PubChem CID 11101
ChEBI CHEBI:66927
IUPAC Name 2-nitrobenzaldehyde
SMILES C1=CC=C(C(=C1)C=O)[N+](=O)[O-]

Specifications

Melting Point 41.0°C to 44.0°C
Color Yellow
Boiling Point 153.0°C (23.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4CHO
Quantity 25 g
Beilstein 07,243
Merck Index 14,6587
Solubility Information Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol,benzene and ether
Formula Weight 151.12
Percent Purity 99+%
Physical Form Crystalline Powder, Chunks or Needles
Chemical Name or Material 2-Nitrobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful to aquatic life with long lasting effects.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for s
EINECSNumber 209-025-3

RUO – Research Use Only