missing translation for 'onlineSavingsMsg'
Learn More

2-Nitroanisole 98.0+%, TCI America™

Catalog No. N012525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N012525G 25 g
N0125500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N012525G Supplier TCI America Supplier No. N012525G

Chemical Identifiers

CAS 91-23-6
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007096
InChI Key CFBYEGUGFPZCNF-UHFFFAOYSA-N
Synonym 2-nitroanisole, o-nitroanisole, anisole, o-nitro, 1-nitro-2-methoxybenzene, 2-methoxynitrobenzene, o-nitrophenyl methyl ether, benzene, 1-methoxy-2-nitro, 2-methoxy-1-nitrobenzene, benzene, methoxynitro, o-nitro methoxy benzene
PubChem CID 7048
ChEBI CHEBI:48722
IUPAC Name 1-methoxy-2-nitrobenzene
SMILES COC1=CC=CC=C1[N+]([O-])=O

Specifications

Color Yellow
Boiling Point 273°C
Quantity 25 g
UN Number 2730
Formula Weight 153.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Nitroanisole
EINECSNumber (3)-0787
RTECSNumber BZ8790000
TSCA Yes