missing translation for 'onlineSavingsMsg'
Learn More

2'-Nitroacetophenone, 97%

CAS: 577-59-3 | C8H7NO3 | 165.15 g/mol

Supplier:  Thermo Scientific Chemicals A1320809

 View more versions of this product

Catalog No. AAA1320809


Only null left
Explore available promotions

Description

Description

2'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. It is also used as a synthetic reagent. It is the sensitizer which can be used as pharmaceutical intermediates. It can also be used to produce 2-nitro-benzoic acid at temperature of 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. It is also used as a synthetic reagent. It is the sensitizer which can be used as pharmaceutical intermediates. It can also be used to produce 2-nitro-benzoic acid at temperature of 20°C.

Solubility
It is Soluble in water, partly in ethanol, ether, chloroform, and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Strong bases.
Specifications

Specifications

2'-Nitroacetophenone
25°C to 28°C
158°C to 160°C (16 mmHg)
Characteristic
1.5515
10 g
2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
SUGXZLKUDLDTKX-UHFFFAOYSA-N
1-(2-nitrophenyl)ethanone
11346
97%
577-59-3
1.24
>110°C (230°F)
C8H7NO3
MFCD00007144
1102322
It is Soluble in water,partly in ethanol,ether,chloroform,and ethyl acetate.
CC(=O)C1=CC=CC=C1[N+]([O-])=O
165.15
165.15

Chemical Identifiers

577-59-3
165.15
SUGXZLKUDLDTKX-UHFFFAOYSA-N
11346
CC(=O)C1=CC=CC=C1[N+]([O-])=O
C8H7NO3
MFCD00007144
2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
1-(2-nitrophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 209-414-8

RTECSNumber : AM9600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only