missing translation for 'onlineSavingsMsg'
Learn More

2'-Nitroacetophenone, 97%

Catalog No. p-7021040
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1320809 10 g
AAA1320818 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1320809 Supplier Thermo Scientific Chemicals Supplier No. A1320809

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 577-59-3 | C8H7NO3 | 165.15 g/mol

2'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. It is also used as a synthetic reagent. It is the sensitizer which can be used as pharmaceutical intermediates. It can also be used to produce 2-nitro-benzoic acid at temperature of 20°C.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. It is also used as a synthetic reagent. It is the sensitizer which can be used as pharmaceutical intermediates. It can also be used to produce 2-nitro-benzoic acid at temperature of 20°C.

Solubility
It is Soluble in water, partly in ethanol, ether, chloroform, and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from Strong oxidizing agents, Strong bases.

Chemical Identifiers

CAS 577-59-3
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007144
InChI Key SUGXZLKUDLDTKX-UHFFFAOYSA-N
Synonym 2'-nitroacetophenone, 1-2-nitrophenyl ethanone, o-nitroacetophenone, ethanone, 1-2-nitrophenyl, 2-acetylnitrobenzene, acetophenone, 2'-nitro, methyl 2-nitrophenyl ketone, unii-52l53k4x3o, ccris 2329, 1-2-nitrophenyl ethan-1-one
PubChem CID 11346
IUPAC Name 1-(2-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 25°C to 28°C
Density 1.24
Boiling Point 158°C to 160°C (16 mmHg)
Flash Point >110°C (230°F)
Odor Characteristic
Refractive Index 1.5515
Quantity 10 g
Beilstein 1102322
Solubility Information It is Soluble in water,partly in ethanol,ether,chloroform,and ethyl acetate.
Formula Weight 165.15
Percent Purity 97%
Chemical Name or Material 2'-Nitroacetophenone
Show More Show Less
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 209-414-8
RTECSNumber AM9600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only