missing translation for 'onlineSavingsMsg'
Learn More

2'-Nitroacetanilide 98.0+%, TCI America™

Catalog No. N010725G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N010725G Supplier TCI America Supplier No. N010725G

Chemical Identifiers

CAS 552-32-9
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00016991
InChI Key BUNFNRVLMKHKIT-UHFFFAOYSA-N
Synonym n-2-nitrophenyl acetamide, 2'-nitroacetanilide, o-nitroacetanilide, 2-nitroacetanilide, acetamide, n-2-nitrophenyl, acetanilide, 2'-nitro, unii-q4kjc83992, o-nitroacetoanilide, zlchem 447, pubchem3357
PubChem CID 11090
IUPAC Name N-(2-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 93°C
Color Yellow
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2′-Nitroacetanilide
EINECSNumber (3)-0396
TSCA Yes