missing translation for 'onlineSavingsMsg'
Learn More

2'-Nitroacetanilide, 98+%

Catalog No. AAA1667706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1667706 5 g
AAA1667714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1667706 Supplier Thermo Scientific Chemicals Supplier No. A1667706

CAS: 552-32-9 | C8H8N2O3 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 552-32-9
Molecular Formula C8H8N2O3
Molecular Weight (g/mol) 180.16
MDL Number MFCD00016991
InChI Key BUNFNRVLMKHKIT-UHFFFAOYSA-N
Synonym n-2-nitrophenyl acetamide, 2'-nitroacetanilide, o-nitroacetanilide, 2-nitroacetanilide, acetamide, n-2-nitrophenyl, acetanilide, 2'-nitro, unii-q4kjc83992, o-nitroacetoanilide, zlchem 447, pubchem3357
PubChem CID 11090
IUPAC Name N-(2-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC=CC=C1[N+]([O-])=O

Specifications

Melting Point 90°C to 93°C
Density 1.42
Quantity 5 g
Beilstein 1959178
Merck Index 14,6581
Formula Weight 180.16
Percent Purity ≥98%
Chemical Name or Material 2'-Nitroacetanilide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 209-009-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only