missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-p-phenylenediamine, 95%

Catalog No. p-7027035
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2347714 25 g
AAB2347722 100 g
AAB2347736 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2347714 Supplier Thermo Scientific Chemicals Supplier No. B2347714
Only null left

CAS: 5307-14-2 | C6H7N3O2 | 153.141 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5307-14-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007903
InChI Key HVHNMNGARPCGGD-UHFFFAOYSA-N
Synonym 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
PubChem CID 4338370
ChEBI CHEBI:76394
IUPAC Name 2-nitrobenzene-1,4-diamine
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])N

Specifications

Melting Point 134°C to 137°C
Quantity 25 g
Formula Weight 153.14
Percent Purity 95%
Chemical Name or Material 2-Nitro-p-phenylenediamine
Hazard Category H341
Hazard Statement GHS H Statement
H341-H351
Suspected of causing genetic defects.
Suspected of causing cancer.
Precautionary Statement P201-P202-P281-P308+P313-P501c
EINECSNumber 226-164-5
RTECSNumber ST3000000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only