accessibility menu, dialog, popup

UserName

NH2NH2NOO

CAS RN 5307-14-2

IUPAC Name: 2-nitrobenzene-1,4-diamine
Synonyms: 1,4-benzenediamine, 2-nitro-
Molecular Weight (g/mol): 153.14
Molecular Formula: C6H7N3O2
InChi Key: HVHNMNGARPCGGD-UHFFFAOYSA-N
SMILES: NC1=CC=C(N)C(=C1)[N+]([O-])=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)