missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-5-(trifluoromethyl)aniline, 97%

Catalog No. AAL1231006
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1231006 5 g
AAL1231003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1231006 Supplier Thermo Scientific Chemicals Supplier No. L1231006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 402-14-2 | C7H5F3N2O2 | 206.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 402-14-2
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.12
MDL Number MFCD00042447
InChI Key AUTLVHYEAAAKNM-UHFFFAOYSA-N
Synonym 2-nitro-5-trifluoromethyl aniline, 3-amino-4-nitrobenzotrifluoride, 3-amino-4-nitrobenzitrifluoride, benzenamine, 2-nitro-5-trifluoromethyl, 5-trifluoromethyl-2-nitrobenzenamine, 2-nitro-5-trifluoromethyl phenylamine, 2-nitro-5-trifluormethyl anilin, pubchem2790, 3-amino-4-nitrobenztrifluoride, 2-nitro-5-trifluoromethyl-aniline
PubChem CID 223100
IUPAC Name 2-nitro-5-(trifluoromethyl)aniline
SMILES NC1=CC(=CC=C1[N+]([O-])=O)C(F)(F)F

Specifications

Melting Point 98°C to 102°C
Quantity 5 g
UN Number UN2811
Beilstein 2732344
Formula Weight 206.13
Percent Purity 97%
Chemical Name or Material 2-Nitro-5-(trifluoromethyl)aniline
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only