missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)phenol 98.0+%, TCI America™

Catalog No. p-7133970
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07791G 1 g
N07795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07791G Supplier TCI America Supplier No. N07791G
Only null left

Chemical Identifiers

CAS 400-99-7
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD00009791
InChI Key XZEDEVRSUANQEM-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl phenol, 4-hydroxy-3-nitrobenzotrifluoride, phenol, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethylphenol, 2-nitro-alpha,alpha,alpha-trifluoro-p-cresol, 4-hydroxy-3-nitrobenzotrifluoroide, p-cresol, 2-nitro-alpha,alpha,alpha-trifluoro, 2-nitro-4-trifluoromethyl-phenol, pubchem4150, 4-06-00-02150 beilstein handbook reference
PubChem CID 9816
IUPAC Name 2-nitro-4-(trifluoromethyl)phenol
SMILES OC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F

Specifications

Boiling Point 94°C
Quantity 1 g
Formula Weight 207.11
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
RTECSNumber GP2984950
TSCA Yes