missing translation for 'onlineSavingsMsg'
Learn More

3-(Benzyldimethylammonio)propanesulfonate, 95%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 428070050

Quantity 5g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC428070050


Only null left

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
Specifications

Chemical Identifiers

81239-45-4
257.35
MEJASPJNLSQOAG-UHFFFAOYSA-N
4386830
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
C12H19NO3S
MFCD00225018
ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate

Specifications

3-(Benzyldimethylammonio)propanesulfonate
94.0
95%
C12H19NO3S
MFCD00225018
Solubility in water: 10g/L (20°C)
C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1
257.35
257.35
81239-45-4
100.0
Glass bottle
5g
ndsb-256, 3-benzyl dimethyl ammonio propane-1-sulfonate, 3-benzyldimethylammonio propane-1-sulfonate, dimethylbenzylammonium propane sulfonate, 3-benzyl dimethyl ammonio propane-1-sulphonate, ndsb 256, dmx, 3-benzyldimethylammonio propanesulfonate, 3-dimethylbenzylamino propanesulfonic acid, 3-benzyl dimethyl azaniumyl propane-1-sulfonate
MEJASPJNLSQOAG-UHFFFAOYSA-N
3-[benzyl(dimethyl)azaniumyl]propane-1-sulfonate
4386830
95%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions