missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)phenol, 98%

Catalog No. AAL1060403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1060403 1 g
AAL1060406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1060403 Supplier Thermo Scientific Chemicals Supplier No. L1060403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 400-99-7 | C7H4F3NO3 | 207.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 400-99-7
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD00009791
InChI Key XZEDEVRSUANQEM-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl phenol, 4-hydroxy-3-nitrobenzotrifluoride, phenol, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethylphenol, 2-nitro-alpha,alpha,alpha-trifluoro-p-cresol, 4-hydroxy-3-nitrobenzotrifluoroide, p-cresol, 2-nitro-alpha,alpha,alpha-trifluoro, 2-nitro-4-trifluoromethyl-phenol, pubchem4150, 4-06-00-02150 beilstein handbook reference
PubChem CID 9816
IUPAC Name 2-nitro-4-(trifluoromethyl)phenol
SMILES OC1=CC=C(C=C1[N+]([O-])=O)C(F)(F)F

Specifications

Density 1.473
Boiling Point 92°C to 94°C (12 mmHg)
Flash Point 95°C (203°F)
Refractive Index 1.504
Quantity 1 g
Beilstein 2215510
Formula Weight 207.11
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)phenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 206-927-9
RTECSNumber GP2984950
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only