Learn More
2-Nitro-4-(trifluoromethyl)aniline, 98%
CAS: 400-98-6 | C7H5F3N2O2 | 206.124 g/mol
Supplier: Thermo Scientific Chemicals A1441318
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2-Nitro-4-(trifluoromethyl)aniline | |
| 105°C to 109°C | |
| MFCD00007155 | |
| UN2811 | |
| 2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride | |
| C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N | |
| 206.124 | |
| 206.13 |
| 400-98-6 | |
| C7H5F3N2O2 | |
| 50 g | |
| 650808 | |
| ATXBGHLILIABGX-UHFFFAOYSA-N | |
| 2-nitro-4-(trifluoromethyl)aniline | |
| 67865 | |
| 98% |
Chemical Identifiers
| 400-98-6 | |
| 206.124 | |
| ATXBGHLILIABGX-UHFFFAOYSA-N | |
| 67865 | |
| C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N |
| C7H5F3N2O2 | |
| MFCD00007155 | |
| 2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride | |
| 2-nitro-4-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319
DOTInformation : Hazard Class: 6.1; Packaging Group: II
EINECSNumber : 206-926-3
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only