missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-4-(trifluoromethyl)aniline, 98%

Catalog No. AAA1441309
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1441309 10 g
AAA1441318 50 g
AAA1441330 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1441309 Supplier Thermo Scientific Chemicals Supplier No. A1441309
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 400-98-6 | C7H5F3N2O2 | 206.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 400-98-6
Molecular Formula C7H5F3N2O2
Molecular Weight (g/mol) 206.124
MDL Number MFCD00007155
InChI Key ATXBGHLILIABGX-UHFFFAOYSA-N
Synonym 2-nitro-4-trifluoromethyl aniline, 4-amino-3-nitrobenzotrifluoride, benzenamine, 2-nitro-4-trifluoromethyl, alpha,alpha,alpha-trifluoro-2-nitro-p-toluidine, 4-trifluoromethyl 2-nitroaniline, 4-amino-3-nitro benzotrifluoride, 4-amino-3-nitrobenzo trifluoride, aniline, 2-nitro-4-trifluoromethyl, 2-nitro-4-trifluoromethyl phenylamine, 3-nitro-4-aminobenzotrifluoride
PubChem CID 67865
IUPAC Name 2-nitro-4-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])N

Specifications

Melting Point 105°C to 109°C
Quantity 10 g
UN Number UN2811
Beilstein 650808
Formula Weight 206.13
Percent Purity 98%
Chemical Name or Material 2-Nitro-4-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: II
EINECSNumber 206-926-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only