missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-1,4-phenylenediamine 94.0+%, TCI America™

Supplier:  TCI America D0105500G

 View more versions of this product

Catalog No. D0105500G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Nitro-1,4-phenylenediamine
138°C
C6H7N3O2
500 g
HVHNMNGARPCGGD-UHFFFAOYSA-N
2-nitrobenzene-1,4-diamine
4338370
153.14
Crystalline Powder
5307-14-2
Black-Green
MFCD00007903
2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
C1=CC(=C(C=C1N)[N+](=O)[O-])N
153.141
CHEBI:76394
≥94.0% (T)

Chemical Identifiers

5307-14-2
153.141
HVHNMNGARPCGGD-UHFFFAOYSA-N
4338370
2-nitrobenzene-1,4-diamine
C6H7N3O2
MFCD00007903
2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
CHEBI:76394
C1=CC(=C(C=C1N)[N+](=O)[O-])N
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0402

RTECSNumber : ST3000000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions