missing translation for 'onlineSavingsMsg'
Learn More

2-Nitro-1,4-phenylenediamine 94.0+%, TCI America™

Catalog No. D010525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D010525G 25 g
D0105500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D010525G Supplier TCI America Supplier No. D010525G

Chemical Identifiers

CAS 5307-14-2
Molecular Formula C6H7N3O2
Molecular Weight (g/mol) 153.141
MDL Number MFCD00007903
InChI Key HVHNMNGARPCGGD-UHFFFAOYSA-N
Synonym 2-nitro-1,4-phenylenediamine, 1,4-diamino-2-nitrobenzene, 2-nitro-p-phenylenediamine, 2,5-diaminonitrobenzene, 1,4-benzenediamine, 2-nitro, durafur brown, 2-nitro-1,4-benzenediamine, o-nitro-p-phenylenediamine, 4-amino-2-nitroaniline, fouramine 2r
PubChem CID 4338370
ChEBI CHEBI:76394
IUPAC Name 2-nitrobenzene-1,4-diamine
SMILES C1=CC(=C(C=C1N)[N+](=O)[O-])N

Specifications

Melting Point 138°C
Color Black-Green
Quantity 25 g
Formula Weight 153.14
Percent Purity ≥94.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Nitro-1,4-phenylenediamine
EINECSNumber (3)-0402
RTECSNumber ST3000000
TSCA Yes