missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyloxyacetic Acid 98.0+%, TCI America™

Catalog No. N0045500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0045500G 500 g
N004525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0045500G Supplier TCI America Supplier No. N0045500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 120-23-0
Molecular Formula C12H10O3
Molecular Weight (g/mol) 202.209
MDL Number MFCD00004066
InChI Key RZCJYMOBWVJQGV-UHFFFAOYSA-N
Synonym 2-naphthoxyacetic acid, 2-naphthyloxy acetic acid, bnoa, 2-naphthyloxyacetic acid, 2-naphthalen-2-yloxy acetic acid, noxa, betoxon, 2-noxa, naphthoxyacetic acid, betapal
PubChem CID 8422
ChEBI CHEBI:50397
IUPAC Name 2-naphthalen-2-yloxyacetic acid
SMILES C1=CC=C2C=C(C=CC2=C1)OCC(=O)O

Specifications

Melting Point 155°C
Quantity 500 g
Formula Weight 202.21
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyloxyacetic Acid
RTECSNumber AI9659000
TSCA Yes