missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthylacetic acid, 99%

CAS: 581-96-4 | C12H10O2 | 186.21 g/mol

$110.50 - $274.73

Chemical Identifiers

CAS 581-96-4
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004126
InChI Key VIBOGIYPPWLDTI-UHFFFAOYSA-N
Synonym 2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid
PubChem CID 11393
ChEBI CHEBI:37837
IUPAC Name 2-naphthalen-2-ylacetic acid
SMILES OC(=O)CC1=CC=C2C=CC=CC2=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC128220050
SDS View Documents
Thermo Scientific Chemicals
128220050
5 g Glass bottle
Each for $110.50
Only null left
 
AC128220250
SDS View Documents
Thermo Scientific Chemicals
128220250
25 g Glass bottle
Each for $274.73
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

581-96-4
186.21
VIBOGIYPPWLDTI-UHFFFAOYSA-N
11393
2-naphthalen-2-ylacetic acid
C12H10O2
MFCD00004126
2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid
CHEBI:37837
OC(=O)CC1=CC=C2C=CC=CC2=C1

Specifications

581-96-4
100.0
White
99%
C12H10O2
MFCD00004126
09, 667
Solubility in water: insoluble
OC(=O)CC1=CC=C2C=CC=CC2=C1
186.21
CHEBI:37837
99%
2-Naphthylacetic acid
98.5
141.0°C to 143.0°C
Authentic
Glass bottle
C10H7CH2CO2H
5 g
2-naphthylacetic acid, 2-naphthaleneacetic acid, 2-naphthalen-2-yl acetic acid, betoxan, 2-2-naphthyl acetic acid, beta-naphthylacetic acid, beta-naphthaleneacetic acid, beta-naphthaleneacetate, naphthalen-2-ylacetic acid, .beta.-naphthaleneacetic acid
VIBOGIYPPWLDTI-UHFFFAOYSA-N
2-naphthalen-2-ylacetic acid
11393
186.21
Crystalline Powder or Flakes
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 209-475-

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only