missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl Trifluoromethanesulfonate 95.0+%, TCI America™

Catalog No. N08451G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08451G 1 g
N08455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N08451G Supplier TCI America Supplier No. N08451G

Chemical Identifiers

CAS 3857-83-8
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192341
InChI Key MDWRQYBWVTXIIJ-UHFFFAOYSA-N
Synonym 2-naphthyl trifluoromethanesulfonate, 2-naphthyl triflate, 2-naphthyltrifluoromethanesulfonate, 2-naphthyltriflate, acmc-1aigh, trifluoromethanesulfonic acid 2-naphthyl ester, methanesulfonic acid, 1,1,1-trifluoro-, 2-naphthalenyl ester, methanesulfonic acid,1,1,1-trifluoro-, 2-naphthalenyl ester
PubChem CID 4546341
IUPAC Name naphthalen-2-yl trifluoromethanesulfonate
SMILES C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 32°C
Color White-Yellow
Boiling Point 87°C
Quantity 1 g
Formula Weight 276.23
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Trifluoromethanesulfonate