missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific™

Catalog No. p-7052704
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5541703 1 g
AAH5541706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5541703 Supplier Thermo Scientific Chemicals Supplier No. H5541703

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 3857-83-8
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192341
InChI Key MDWRQYBWVTXIIJ-UHFFFAOYSA-N
Synonym 2-naphthyl trifluoromethanesulfonate, 2-naphthyl triflate, 2-naphthyltrifluoromethanesulfonate, 2-naphthyltriflate, acmc-1aigh, trifluoromethanesulfonic acid 2-naphthyl ester, methanesulfonic acid, 1,1,1-trifluoro-, 2-naphthalenyl ester, methanesulfonic acid,1,1,1-trifluoro-, 2-naphthalenyl ester
PubChem CID 4546341
IUPAC Name naphthalen-2-yl trifluoromethanesulfonate
SMILES C1=CC=C2C=C(C=CC2=C1)OS(=O)(=O)C(F)(F)F

Specifications

Melting Point 30°C to 32°C
Color Orange to White
Flash Point 113°C (235°F)
Quantity 1 g
UN Number UN3261
Formula Weight 276.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Naphthyl trifluoromethanesulfonate
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures