missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthyl Benzoate 98.0+%, TCI America™
$57.79 - $57.79
Chemical Identifiers
| CAS | 93-44-7 |
|---|---|
| Molecular Formula | C17H12O2 |
| Molecular Weight (g/mol) | 248.281 |
| MDL Number | MFCD00014320 |
| InChI Key | DWJIJRSTYFPKGD-UHFFFAOYSA-N |
| Synonym | 2-naphthyl benzoate, benzonaphthol, 2-naphthalenol, benzoate, betabenzon, 2-benzoyloxynaphthalene, 2-naphthylbenzoate, 2-naphthol benzoate, beta-naphthol benzoate, beta-naphthyl benzoate, benzoic acid 2-naphthyl ester |
| PubChem CID | 66734 |
| IUPAC Name | naphthalen-2-yl benzoate |
| SMILES | C1=CC=C(C=C1)C(=O)OC2=CC3=CC=CC=C3C=C2 |
Chemical Identifiers
| 93-44-7 | |
| 248.281 | |
| DWJIJRSTYFPKGD-UHFFFAOYSA-N | |
| 66734 | |
| C1=CC=C(C=C1)C(=O)OC2=CC3=CC=CC=C3C=C2 |
| C17H12O2 | |
| MFCD00014320 | |
| 2-naphthyl benzoate, benzonaphthol, 2-naphthalenol, benzoate, betabenzon, 2-benzoyloxynaphthalene, 2-naphthylbenzoate, 2-naphthol benzoate, beta-naphthol benzoate, beta-naphthyl benzoate, benzoic acid 2-naphthyl ester | |
| naphthalen-2-yl benzoate |
Specifications
| 93-44-7 | |
| C17H12O2 | |
| 25 g | |
| DWJIJRSTYFPKGD-UHFFFAOYSA-N | |
| naphthalen-2-yl benzoate | |
| 66734 | |
| ≥98.0% (GC) | |
| 2-Naphthyl Benzoate |
| 109°C | |
| MFCD00014320 | |
| 2-naphthyl benzoate, benzonaphthol, 2-naphthalenol, benzoate, betabenzon, 2-benzoyloxynaphthalene, 2-naphthylbenzoate, 2-naphthol benzoate, beta-naphthol benzoate, beta-naphthyl benzoate, benzoic acid 2-naphthyl ester | |
| C1=CC=C(C=C1)C(=O)OC2=CC3=CC=CC=C3C=C2 | |
| 248.281 | |
| 248.28 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (4)-0358
TSCA : Yes