missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl Acetate 98.0+%, TCI America™

Catalog No. A003825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A003825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A003825G Supplier TCI America Supplier No. A003825G

Chemical Identifiers

CAS 1523-11-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004060
InChI Key RJNPPEUAJCEUPV-UHFFFAOYSA-N
Synonym 2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
PubChem CID 73709
IUPAC Name naphthalen-2-yl acetate
SMILES CC(=O)OC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 70°C
Color White-Yellow
Boiling Point 132°C
Quantity 25 g
Formula Weight 186.21
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthyl Acetate