accessibility menu, dialog, popup

UserName

H3COO

CAS RN 1523-11-1

IUPAC Name: naphthalen-2-yl acetate
Synonyms: naphthalen-2-yl acetate
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: RJNPPEUAJCEUPV-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)