missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl acetate, 99%

CAS: 1523-11-1 | C12H10O2 | 186.21 g/mol

$59.66 - $660.22

Chemical Identifiers

CAS 1523-11-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004060
InChI Key RJNPPEUAJCEUPV-UHFFFAOYSA-N
Synonym 2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
PubChem CID 73709
IUPAC Name naphthalen-2-yl acetate
SMILES CC(=O)OC1=CC=C2C=CC=CC2=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1848709
View Documents
Thermo Scientific Chemicals
A1848709
10 g
Each for $59.66
Only null left
 
AAA1848718
View Documents
Thermo Scientific Chemicals
A1848718
50 g
Each for $169.63
Only null left
 
AAA1848730
View Documents
Thermo Scientific Chemicals
A1848730
250 g
Each for $660.22
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1523-11-1
186.21
RJNPPEUAJCEUPV-UHFFFAOYSA-N
73709
CC(=O)OC1=CC=C2C=CC=CC2=C1
C12H10O2
MFCD00004060
2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
naphthalen-2-yl acetate

Specifications

1523-11-1
C12H10O2
10 g
2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
CC(=O)OC1=CC=C2C=CC=CC2=C1
186.21
186.21
2-Naphthyl acetate
68°C to 70°C
MFCD00004060
1868291
RJNPPEUAJCEUPV-UHFFFAOYSA-N
naphthalen-2-yl acetate
73709
99%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-194-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only