missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthyl acetate, 99%

Catalog No. AAA1848709
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1848709 10 g
AAA1848718 50 g
AAA1848730 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1848709 Supplier Thermo Scientific Chemicals Supplier No. A1848709

CAS: 1523-11-1 | C12H10O2 | 186.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1523-11-1
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00004060
InChI Key RJNPPEUAJCEUPV-UHFFFAOYSA-N
Synonym 2-naphthyl acetate, 2-acetoxynaphthalene, 2-naphthalenol, acetate, beta-naphthyl acetate, 2-naphthol, acetate, naphthalene, 2-acetoxy, 2-naphthalenol, 2-acetate, .beta.-naphthol acetate, .beta.-naphthyl acetate, o-acetyl-.beta.-naphthol
PubChem CID 73709
IUPAC Name naphthalen-2-yl acetate
SMILES CC(=O)OC1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 68°C to 70°C
Quantity 10 g
Beilstein 1868291
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 2-Naphthyl acetate
EINECSNumber 216-194-7
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only