missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoyl chloride, 98%

CAS: 2243-83-6 | C11H7ClO | 190.626 g/mol

$118.99 - $1,991.26

Chemical Identifiers

CAS 2243-83-6
Molecular Formula C11H7ClO
Molecular Weight (g/mol) 190.626
MDL Number MFCD00004093
InChI Key XNLBCXGRQWUJLU-UHFFFAOYSA-N
Synonym 2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
PubChem CID 75246
IUPAC Name naphthalene-2-carbonyl chloride
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1404609
View Documents
Thermo Scientific Chemicals
A1404609
10 g N/A
Only null left
 
AAA1404618
View Documents
Thermo Scientific Chemicals
A1404618
50 g N/A
Only null left
 
AAA1404630
View Documents
Thermo Scientific Chemicals
A1404630
250 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2243-83-6
190.626
XNLBCXGRQWUJLU-UHFFFAOYSA-N
75246
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl
C11H7ClO
MFCD00004093
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
naphthalene-2-carbonyl chloride

Specifications

2243-83-6
159°C to 160°C (9 mmHg)
Pungent
MFCD00004093
UN3261
Moisture sensitive
XNLBCXGRQWUJLU-UHFFFAOYSA-N
naphthalene-2-carbonyl chloride
75246
98%
48°C to 52°C
>110°C (230°F)
C11H7ClO
10 g
907776
2-naphthoyl chloride, 2-naphthalenecarbonyl chloride, 2-naphthoic chloride, 2-naphthoylchloride, beta-naphthoyl chloride, 2-napthoyl chloride, 2-chlorocarbonyl naphthalene, .beta.-naphthoyl chloride, .beta.-naphthalenecarbonyl chloride, beta-naphthalenecarbonyl chloride
C1=CC=C2C=C(C=CC2=C1)C(=O)Cl
190.626
190.63
2-Naphthoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 218-822-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only