missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 156970050

Catalog No. AC156970050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Naphthol
97.5
285.0°C to 286.0°C
Authentic
Plastic drum
C10H7OH
5kg
15,6469
Solubility in water: 1g/L (20°C). Other solubilities: 1 g/80mL boiling water, 1 g/0.8mL alcohol, 1 g/17mL chloroform, 1 g/1.3mL ether, soluble in ethanol, ether, chloroform, glycerol
C1=CC=C2C=C(C=CC2=C1)O
144.17
CHEBI:10432
98%
135-19-3
100.0
153°C
97.5% min. (GC)
C10H8O
MFCD00004067
06,627
2-naphthol, 2-naphthalenol, beta-naphthol, 2-hydroxynaphthalene, betanaphthol, isonaphthol, developer bn, naphthol b, azogen developer a, beta-napthol
JWAZRIHNYRIHIV-UHFFFAOYSA-N
naphthalen-2-ol
8663
144.17
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Very toxic to aquatic life.

GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning

EINECSNumber : 205-182-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only