missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Naphthoic Acid 98.0+%, TCI America™
Supplier: TCI America N00255G
Specifications
| 2-Naphthoic Acid | |
| 185°C | |
| C11H8O2 | |
| 5 g | |
| UOBYKYZJUGYBDK-UHFFFAOYSA-N | |
| naphthalene-2-carboxylic acid | |
| 7123 | |
| 172.18 | |
| Crystalline Powder |
| 93-09-4 | |
| White-Yellow | |
| MFCD00004101 | |
| 2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid | |
| C1=CC=C2C=C(C=CC2=C1)C(=O)O | |
| 172.183 | |
| CHEBI:36106 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 93-09-4 | |
| 172.183 | |
| UOBYKYZJUGYBDK-UHFFFAOYSA-N | |
| 7123 | |
| naphthalene-2-carboxylic acid |
| C11H8O2 | |
| MFCD00004101 | |
| 2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid | |
| CHEBI:36106 | |
| C1=CC=C2C=C(C=CC2=C1)C(=O)O |
Safety and Handling
EINECSNumber : (4)-0397
RTECSNumber : QL1050000
TSCA : Yes