missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthoic acid, 98+%

Catalog No. AAL0510214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0510214 25 g
AAL0510206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0510214 Supplier Thermo Scientific Chemicals Supplier No. L0510214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 93-09-4 | C11H8O2 | 172.183 g/mol

2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Naphthoic acid is an unprotected acid undergoes lithiation, e.g. with s-BuLi, by stereospecific 1,4-addition, providing a facile route to 1,2,2-trisubstituted 1,2-dihydronaphthalenes. It is also used as an intermediate in organic synthesis. The fluorescence spectra and electronic absorption of 2-naphthoic acid was studied.

Solubility
Soluble in alcohol, diethyl ether, methanol, and ethyl acetate (20 mg/ml). Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.

Chemical Identifiers

CAS 93-09-4
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.183
MDL Number MFCD00004101
InChI Key UOBYKYZJUGYBDK-UHFFFAOYSA-N
Synonym 2-naphthoic acid, 2-naphthalenecarboxylic acid, isonaphthoic acid, 2-carboxynaphthalene, 2-maythic acid, beta-naphthoic acid, ne-2-carboxylic acid, unii-qlg01v0w2l, naphthalene-beta-carboxylic acid, .beta.-naphthoic acid
PubChem CID 7123
ChEBI CHEBI:36106
IUPAC Name naphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)O

Specifications

Melting Point 182°C to 186°C
Density 1.08
Flash Point 205°C (401°F)
Quantity 25 g
Beilstein 972039
Merck Index 14,6382
Solubility Information Soluble in alcohol,diethyl ether,methanol,and ethyl acetate (20mg/ml). Insoluble in water.
Formula Weight 172.18
Percent Purity ≥98%
Chemical Name or Material 2-Naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-217-8
RTECSNumber QL1050000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only