missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenesulfonyl Chloride 98.0+%, TCI America™

$86.64 - $86.64

Chemical Identifiers

CAS 93-11-8
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.67
MDL Number MFCD00004087
InChI Key OPECTNGATDYLSS-UHFFFAOYSA-N
Synonym 2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
PubChem CID 7125
IUPAC Name naphthalene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N001825G
View Documents
TCI America
N001825G
25 g
Each for $86.64
Only null left
 
Description

Description

Specifications

Chemical Identifiers

93-11-8
226.67
OPECTNGATDYLSS-UHFFFAOYSA-N
7125
ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
C10H7ClO2S
MFCD00004087
2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
naphthalene-2-sulfonyl chloride

Specifications

93-11-8
White-Yellow
MFCD00004087
3261
OPECTNGATDYLSS-UHFFFAOYSA-N
naphthalene-2-sulfonyl chloride
7125
≥98.0% (GC,T)
2-Naphthalenesulfonyl Chloride
77°C
C10H7ClO2S
25 g
2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1
226.67
226.67
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents