missing translation for 'onlineSavingsMsg'
Learn More

2-Naphthalenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. N001825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N001825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N001825G Supplier TCI America Supplier No. N001825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 93-11-8
Molecular Formula C10H7ClO2S
Molecular Weight (g/mol) 226.67
MDL Number MFCD00004087
InChI Key OPECTNGATDYLSS-UHFFFAOYSA-N
Synonym 2-naphthalenesulfonyl chloride, 2-naphthylsulfonyl chloride, beta-naphthalenesulfonyl chloride, naphthalene-2-sulfonic acid chloride, .beta.-naphthalenesulfochloride, naphthalene-2-sulphonyl chloride, 2-naphthalenesulfonylchloride, 2-naphthalenylsulfonyl chloride, 2-naphthalene sulfonyl chloride, .beta.-naphthalenesulfonyl chloride
PubChem CID 7125
IUPAC Name naphthalene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C2C=CC=CC2=C1

Specifications

Melting Point 77°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 226.67
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Naphthalenesulfonyl Chloride