missing translation for 'onlineSavingsMsg'
Learn More

2-n-Propyl-1,3-propanediol, 98%

Catalog No. AAB2215203
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2215203 1 g
AAB2215206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2215203 Supplier Thermo Scientific Chemicals Supplier No. B2215203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2612-28-4 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2612-28-4
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00799394
InChI Key FZHZPYGRGQZBCV-UHFFFAOYSA-N
Synonym 2-n-propylpropane-1,3-diol, 1,3-propanediol, 2-propyl, 2-propyl-1,3-propanediol, 2-n-propyl-1,3-propanediol, 2-propylpropan-1,3-diol, 2-propyl-propane-l,3-diol, 1,3-propanediol,2-propyl, 1,1-bis hydroxymethyl butane, 2-n-propyl-propane-1,3-diol
PubChem CID 7021156
IUPAC Name 2-propylpropane-1,3-diol
SMILES CCCC(CO)CO

Specifications

Quantity 1 g
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material 2-n-Propyl-1,3-propanediol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only