accessibility menu, dialog, popup

UserName

CAS RN 2612-28-4

IUPAC Name: 2-propylpropane-1,3-diol
Synonyms: 2-propylpropane-1,3-diol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: FZHZPYGRGQZBCV-UHFFFAOYSA-N
SMILES: CCCC(CO)CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results