missing translation for 'onlineSavingsMsg'
Learn More

2-n-Pentylpropane-1,3-diol, 97%, Thermo Scientific Chemicals

Catalog No. AAB2180006
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2180006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAB2180006 Supplier Thermo Scientific Chemicals Supplier No. B2180006
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25462-23-1
Molecular Formula C8H18O2
Molecular Weight (g/mol) 146.23
MDL Number MFCD00801123
InChI Key RHYUFGNCUXTFTC-UHFFFAOYSA-N
Synonym 2-pentyl-1,3-propanediol, 1,3-propanediol, 2-pentyl, 2-n-pentylpropane-1,3-diol, 2-pentylpropane-1,3diol, 2-n-pentylpropane-1,3diol, 2-n-pentylpropan-1,3-diol, 2-n-pentyl-1,3-propanediol, 2-n-pentylpropane-1, 3-diol
PubChem CID 117509
IUPAC Name 2-pentylpropane-1,3-diol
SMILES CCCCCC(CO)CO

Specifications

Quantity 5 g
Formula Weight 146.23
Percent Purity 97%
Chemical Name or Material 2-n-Pentylpropane-1,3-diol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only